2-Bromofluorene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS21478G100EA 0 CHF
ACTIAS21478G100 ACTIAS21478G25
2-Bromofluorene
2-Bromofluorene
Formula: C₁₃H₉Br
MW: 245.12 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 102…103 °C
MDL Number: MFCD00001115
CAS Number: 1133-80-8
EINECS: 214-480-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR