1-Methylcyclopentanol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21483G5EA 0 CHF
ACTIAS21483G5 ACTIAS21483G25
1-Methylcyclopentanol
1-Methylcyclopentanol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: 32…33 °C
Density: 0.905 g/cm³ (23.5 °C)
MDL Number: MFCD00001364
CAS Number: 1462-03-9
EINECS: 215-963-4
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR