5-Methyl-1,3-cyclohexandion
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS35452G5EA
0
CHF
ACTIAS35452G5
ACTIAS35452G1
5-Methyl-1,3-cyclohexandion
5-Methyl-1,3-cyclohexandion
Formel:
C₇H₁₀O₂ MW: 126,16 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00010379 CAS-Nummer: 4341-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...