2-Amino-3,5-dichloro-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS4093G5EA 0 CHF
ACTIAS4093G5 ACTIAS4093G25
2-Amino-3,5-dichloro-6-methylpyridine
2-Amino-3,5-dichloro-6-methylpyridine
Formula: C₆H₆Cl₂N₂
MDL Number: MFCD00129029
CAS Number: 22137-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR