3-Amino-2-bromo-4-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Amino-2-bromo-4-picoline

ACTIAS33404G1EA 0 CHF
ACTIAS33404G1 ACTIAS33404G10 ACTIAS33404G5
3-Amino-2-bromo-4-methylpyridine
3-Amino-2-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD04112497
CAS Number: 126325-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR