2-Acetamido-3-brom-5-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32107G1EA 0 CHF
ACTIAS32107G1 ACTIAS32107G5
2-Acetamido-3-brom-5-fluorpyridin
2-Acetamido-3-brom-5-fluorpyridin
Formel: C₇H₆BrFN₂O
MDL Number: MFCD11053869
CAS-Nummer: 1065074-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR