Methyl-2-brom-5-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS25279G25EA 51 CHF
ACTIAS25279G25
Methyl-2-brom-5-fluorbenzoat
Methyl-2-brom-5-fluorbenzoat
Formel: C₈H₆BrFO₂
MW: 233,04 g/mol
MDL Number: MFCD00086167
CAS-Nummer: 6942-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR