Samarium (III) trifluoromethanesulphonate

Synonyms: Samarium(III) trifluoromethanesulphonate

ACTIAS23484G25EA 0 CHF
ACTIAS23484G25
Samarium (III) trifluoromethanesulphonate
Samarium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
MDL Number: MFCD00192431
CAS Number: 52093-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR