Cerium (III) trifluoromethanesulphonate

ACTIAS23549G100EA 0 CHF
ACTIAS23549G100 ACTIAS23549G25
Cerium (III) trifluoromethanesulphonate
Cerium (III) trifluoromethanesulphonate
Formula: C₃CeF₉O₉S₃
MDL Number: MFCD02093899
CAS Number: 76089-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR