4-(Boc-amino)-2-chloropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Amino-2-chloropyridine, 4-BOC protected

ACTIAS12845G5EA 0 CHF
ACTIAS12845G25 ACTIAS12845G5
4-(Boc-amino)-2-chloropyridine
4-(Boc-amino)-2-chloropyridine
Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD02179232
CAS Number: 234108-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR