DL-α-Aminoadipic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
H-DL-Aad-OH
, H-DL-Homoglu-OH
, DL-2-Aminoadipic acid
, (RS)-2-Aminohexanedioic acid
ACTIAS35200G5EA
0
CHF
ACTIAS35200G5
ACTIAS35200G1
ACTIAS35200G10
DL-α-Aminoadipic acid
DL-α-Aminoadipic acid
Formula:
C₆H₁₁NO₄ |
MDL Number:
MFCD00063119 CAS Number: 542-32-5 EINECS: 208-809-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...