5-Bromo-2-furoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromofuroic acid

ACTIAS7122G100EA 0 CHF
ACTIAS7122G100 ACTIAS7122G25
5-Bromo-2-furoic acid
5-Bromo-2-furoic acid
Formula: C₅H₃BrO₃
MW: 190.98 g/mol
MDL Number: MFCD00003239
CAS Number: 585-70-6
EINECS: 209-559-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR