4-Bromosalicylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Bromo-2-hydroxybenzoic acid

ACTIAS21314G25EA 75 CHF
ACTIAS21314G25
4-Bromosalicylic acid
4-Bromosalicylic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD06797970
CAS Number: 1666-28-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR