4-Chlorpyridin Hydrochlorid

ACTIAS20134G100EA 0 CHF
ACTIAS20134G100
4-Chlorpyridin Hydrochlorid
4-Chlorpyridin Hydrochlorid
Formel: C₅H₅Cl₂N
MDL Number: MFCD00012829
CAS-Nummer: 7379-35-3
EINECS: 230-946-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR