Copper(II) trifluoromethanesulphonate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Copper triflateCopper(II) trifluoromethanesulfonate

ACTIAS23138G100EA 0 CHF
ACTIAS23138G5 ACTIAS23138G100 ACTIAS23138G25
Copper(II) trifluoromethanesulphonate
Copper (II) trifluoromethanesulphonate
Formula: C₂CuF₆O₆S₂
MW: 361.69 g/mol
MDL Number: MFCD00077492
CAS Number: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR