1-(4-tert-Butylbenzyl)piperazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24299G25EA 108 CHF
ACTIAS24299G25 ACTIAS24299G100
1-(4-tert-Butylbenzyl)piperazine
1-(4-tert-Butylbenzyl)piperazine
Formula: C₁₅H₂₄N₂
MW: 232.37 g/mol
MDL Number: MFCD00082594
CAS Number: 956-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR