1-Aminocyclohexanecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Amino-1-cyclohexanecarboxylic acid1-Aminocyclohexane-1-carboxylic acid

ACTIAS20415G100EA 457 CHF
ACTIAS20415G100
1-Aminocyclohexanecarboxylic acid
1-Aminocyclohexanecarboxylic acid
Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 170 °C
Density: 1.229 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001487
CAS Number: 2756-85-6
EINECS: 220-411-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR