8-Methylquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS23884G100EA 303 CHF
ACTIAS23884G25 ACTIAS23884G100
8-Methylquinoline
8-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1,0852 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006810
CAS Number: 611-32-5
EINECS: 210-264-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR