7-Methylquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35367G1EA 31 CHF
ACTIAS35367G1 ACTIAS35367G5 ACTIAS35367G10
7-Methylquinoline
7-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 39 °C
Density: 1,084 g/cm³ (0 °C)
MDL Number: MFCD00006805
CAS Number: 612-60-2
EINECS: 210-316-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR