4-Methyl-2-quinolinol

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 4-Methylquinolin-2-ol2-Lepidinol2-Hydroxy-4-methylquinoline

ACTIAS34849G5EA 0 CHF
ACTIAS34849G1 ACTIAS34849G5
4-Methyl-2-quinolinol
4-Methyl-2-quinolinol
Formula: C₁₀H₉NO
MW: 159.185 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 220…222 °C
MDL Number: MFCD00006745
CAS Number: 607-66-9
EINECS: 210-139-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR