6-Nitroquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31913G100EA 0 CHF
ACTIAS31913G100
6-Nitroquinoline
6-Nitroquinoline
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 75 °C
MDL Number: MFCD00006799
CAS Number: 613-50-3
EINECS: 210-346-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR