4-Chloroquinoline
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS1632G5EA
0
CHF
ACTIAS1632G5
ACTIAS1632G1
ACTIAS1632G25
4-Chloroquinoline
4-Chloroquinoline
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 260…261 °C (1013 hPa) Melting Pt: 28 °C Density: 1,25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 EINECS: 210-267-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...