2-Chloroquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS7146G5EA 0 CHF
ACTIAS7146G5 ACTIAS7146G25
2-Chloroquinoline
2-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 266…267 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1,2464 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006741
CAS Number: 612-62-4
EINECS: 210-317-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR