5-Bromoquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS1006G5EA 0 CHF
ACTIAS1006G5 ACTIAS1006G25
5-Bromoquinoline
5-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
MDL Number: MFCD00234481
CAS Number: 4964-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR