4-Bromoquinoline
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Danger
|
ACTIAS1343G5EA
0
CHF
ACTIAS1343G5
ACTIAS1343G1
ACTIAS1343G25
4-Bromoquinoline
4-Bromoquinoline
Formula:
C₉H₆BrN MW: 208.06 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07644514 CAS Number: 3964-04-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...