1,3,5-Tri-tert-butylbenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35125G5EA 31 CHF
ACTIAS35125G10 ACTIAS35125G1 ACTIAS35125G5
1,3,5-Tri-tert-butylbenzene
1,3,5-Tri-tert-butylbenzene
Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 69…71 °C
Density: 2,637 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008831
CAS Number: 1460-02-2
EINECS: 215-952-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR