3,5-Dimethylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40290G100EA 0 CHF
ACTIAS40290G25 ACTIAS40290G100
3,5-Dimethylbenzoic acid
3,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 166…169 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR