2,6-Dimethylbenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32449G5EA 0 CHF
ACTIAS32449G5 ACTIAS32449G10
2,6-Dimethylbenzonitrile
2,6-Dimethylbenzonitrile
Formula: C₉H₉N
MW: 131.18 g/mol
MDL Number: MFCD00037166
CAS Number: 6575-13-9
EINECS: 229-503-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR