2-Brom-6-methoxynaphthalin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS40191G100EA
0
CHF
ACTIAS40191G100
2-Brom-6-methoxynaphthalin
2-Bromo-6-methoxynaphthalene
Formel:
C₁₁H₉BrO MW: 237,1 g/mol |
MDL Number:
MFCD00004062 CAS-Nummer: 5111-65-9 EINECS: 225-837-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...