4-Methyl-3-thiopheneboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32291G1EA 0 CHF
ACTIAS32291G1
4-Methyl-3-thiopheneboronic acid
4-Methyl-3-thiopheneboronic acid
Formula: C₅H₇BO₂S
MW: 141.99 g/mol
MDL Number: MFCD04039882
CAS Number: 177735-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR