2-Formyl-3-thiophenboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS51450G5EA
0
CHF
ACTIAS51450G5
ACTIAS51450G1
2-Formyl-3-thiophenboronsäure
2-Formyl-3-thiophenboronsäure
Formel:
C₅H₅BO₃S MW: 155,97 g/mol |
MDL Number:
MFCD01075678 CAS-Nummer: 4347-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...