4-Morpholinoanilin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20448G25EA 0 CHF
ACTIAS20448G25
4-Morpholinoanilin
4-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
MW: 178,23 g/mol
MDL Number: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR