(R)-(+)-4-Brom-α-methylbenzylamin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS23080G5EA
0
CHF
ACTIAS23080G5
ACTIAS23080G25
(R)-(+)-4-Brom-α-methylbenzylamin
(R)-(+)-4-Brom-α-methylbenzylamin
Formel:
C₈H₁₀BrN MW: 200,08 g/mol |
MDL Number:
MFCD00025548 CAS-Nummer: 45791-36-4 EINECS: 256-245-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...