4-Nitrophenethylbromid

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS23099G25EA 24 CHF
ACTIAS23099G25
4-Nitrophenethylbromid
4-Nitrophenethylbromid
Formel: C₈H₈BrNO₂
MW: 230,06 g/mol
MDL Number: MFCD00007386
CAS-Nummer: 5339-26-4
EINECS: 226-271-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR