4-Hydroxy-3,5-dimethylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 3,5-Dimethyl-4-hydroxybenzaldehyde

ACTIAS40286G100EA 0 CHF
ACTIAS40286G25 ACTIAS40286G100
4-Hydroxy-3,5-dimethylbenzaldehyde
4-Hydroxy-3,5-dimethylbenzaldehyde
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006946
CAS Number: 2233-18-3
EINECS: 218-774-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR