4-Fluorophenethylamine

ACTIAS21084G100EA 0 CHF
ACTIAS21084G100
4-Fluorophenethylamine
4-Fluorophenethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD00134208
CAS Number: 1583-88-6
EINECS: 216-435-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR