3-Fluorophenethylamine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-(3-Fluorophenyl)ethylamine

ACTIAS20572G25EA 0 CHF
ACTIAS20572G25 ACTIAS20572G100
3-Fluorophenethylamine
3-Fluorophenethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD00075376
CAS Number: 404-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR