2-Fluorophenethylamine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-(2-Fluorophenyl)ethylamine2-Fluorophenethylamine1-Amino-2-(2-fluorophenyl)ethane

ACTIAS20571G25EA 0 CHF
ACTIAS20571G25 ACTIAS20571G100
2-Fluorophenethylamine
2-Fluorophenethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD00075502
CAS Number: 52721-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR