3-Chlorophenethylamine

Synonyms: 2-(3-Chlorophenyl)ethylamine

ACTIAS23508G100EA 0 CHF
ACTIAS23508G100 ACTIAS23508G25
3-Chlorophenethylamine
3-Chlorophenethylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
MDL Number: MFCD00047957
CAS Number: 13078-79-0
EINECS: 235-982-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR