3,4-Dichlorophenethylamine
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS24938G5EA
0
CHF
ACTIAS24938G5
ACTIAS24938G25
ACTIAS24938G100
3,4-Dichlorophenethylamine
3,4-Dichlorophenethylamine
Formula:
C₈H₉Cl₂N MW: 190.07 g/mol |
MDL Number:
MFCD00060617 CAS Number: 21581-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...