2-Chlorophenethylamine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24054G100EA 0 CHF
ACTIAS24054G100 ACTIAS24054G25
2-Chlorophenethylamine
2-Chlorophenethylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
MDL Number: MFCD00008185
CAS Number: 13078-80-3
EINECS: 235-982-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR