2-Bromphenethylamin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS32973G5EA
0
CHF
ACTIAS32973G5
ACTIAS32973G1
2-Bromphenethylamin
2-Bromphenethylamin
Formel:
C₈H₁₀BrN MW: 200,08 g/mol |
MDL Number:
MFCD01529874 CAS-Nummer: 65185-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...