3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40296G100EA 0 CHF
ACTIAS40296G10 ACTIAS40296G100
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 190 °C
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR