4-Fluoro-2-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS21014G100EA 0 CHF
ACTIAS21014G100
4-Fluoro-2-nitrotoluene
4-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 27 °C
Density: 1.2911 g/cm³ (0 °C)
MDL Number: MFCD00007200
CAS Number: 446-10-6
EINECS: 207-161-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR