4-Hydroxy-3-methylbenzoic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
4-Hydroxy-m-toluic acid
ACTIAS20538G25EA
46
CHF
ACTIAS20538G25
ACTIAS20538G100
4-Hydroxy-3-methylbenzoic acid
4-Hydroxy-3-methylbenzoic acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol |
MDL Number:
MFCD00270105 CAS Number: 499-76-3 EINECS: 207-890-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...