2-Hydroxy-p-toluic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
Synonyms:
4-Methylsalicylic acid
, m-Cresotic acid
, m-Homosalicyclic acid
, 2-Hydroxy-4-methylbenzoic acid
ACTIAS34846G100EA
0
CHF
ACTIAS34846G100
ACTIAS34846G10
2-Hydroxy-p-toluic acid
2-Hydroxy-p-toluic acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 177 °C |
MDL Number:
MFCD00002454 CAS Number: 50-85-1 EINECS: 200-068-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...