5-Nitro-o-toluic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Methyl-5-nitrobenzoic acid

ACTIAS1511G100EA 0 CHF
ACTIAS1511G100 ACTIAS1511G25
5-Nitro-o-toluic acid
5-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 179…181 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR