4-Isopropylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS20393G100EA 0 CHF
ACTIAS20393G100 ACTIAS20393G10 ACTIAS20393G25 ACTIAS20393G5
4-Isopropylbenzoesäure
4-Isopropylbenzoesäure
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Siedepunkt: 272 °C (1013 hPa)
Melting Pt: 115…117 °C
Dichte: 1,162 g/cm³ (20 °C)
MDL Number: MFCD00002564
CAS-Nummer: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR