α-Bromo-2-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 1-(Bromomethyl)-2-nitrobenzenealpha-Bromo-2-nitrotoluene2-Nitrobenzyl bromideα-Bromo-o-nitrotoluene

ACTIAS40258G100EA 0 CHF
ACTIAS40258G100
α-Bromo-2-nitrotoluene
α-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR