4-Brom-3-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Amino-2-brom-α,α,α-trifluortoluol4-Brom-α,α,α-trifluor-m-toluidin

ACTIAS40400G100EA 0 CHF
ACTIAS40400G100
4-Brom-3-(trifluormethyl)anilin
4-Brom-3-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MW: 240,02 g/mol
Siedepunkt: 81…84 °C (0,5 mmHg)
Melting Pt: 55…56 °C
MDL Number: MFCD00007827
CAS-Nummer: 393-36-2
EINECS: 206-885-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR